Ammonium hexafluorosilicate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate

128075-1KG 128075-250G 128075-5KG
BT128075-1KGEA 85 USD
BT128075-1KG BT128075-250G BT128075-5KG
Ammonium hexafluorosilicate ≥98%
Ammonium hexafluorosilicate

CAS: 16919-19-0; EC No: 240-968-3; MDL No: MFCD00010887 UN No: UN2854; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: (NH4)2SiF6 ; MW: 178.15 Density (g/mL): 2.01


  • Powder

Formula: H₈F₆N₂Si
Density: 2.01 g/mL
MDL Number: MFCD00010887
CAS Number: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR