1-(2,4-Difluorophenyl)piperazine ≥99%

Supplier: BeanTown Chemical

128040-1G 128040-25G 128040-5G
BT128040-1GEA 38 USD
BT128040-1G BT128040-25G BT128040-5G
1-(2,4-Difluorophenyl)piperazine ≥99%
1-(2,4-Difluorophenyl)piperazine

CAS: 115761-79-0; MDL No: MFCD00082588 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II Crystalline/Powder; Molecular Formula: C10H12F2N2; MW: 198.22 Air Sensitive


  • Air Sensitive
  • Crystalline/Powder

Formula: C₁₀H₁₂F₂N₂
MDL Number: MFCD00082588
CAS Number: 115761-79-0
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR