Sodium dehydracetate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Dehydracetic acid sodium salt, Dehydroacetic acid sodium salt, Sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide, Sodium dehydroacetate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate
128020-100G
128020-2.5KG
128020-500G
BT128020-100GEA
34
USD
BT128020-100G
BT128020-2.5KG
BT128020-500G
Sodium dehydracetate ≥97%
Sodium dehydracetate
CAS: 4418-26-2; EC No: 224-580-1; MDL No: MFCD00040583; RTECS: UP8225000
Powder; Molecular Formula: C8H7NaO4; MW: 190.13
Melting Point: 295° (decomposes)
- RTECS: UP8225000
- Powder
Formula:
C₈H₇NaO₄ Melting Pt: 295 °C (dec.) |
MDL Number:
MFCD00040583 CAS Number: 4418-26-2 EINECS: 224-580-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...