2,2'-Bipyridyl ≥98.0% ACS
Supplier: BeanTown Chemical
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
127950-10G
127950-250G
127950-50G
BT127950-10GEA
28.26
USD
BT127950-10G
BT127950-250G
BT127950-50G
2,2'-Bipyridyl ≥98.0% ACS
2,2'-Bipyridyl
CAS: 366-18-7; EC No: 206-674-4; MDL No: MFCD00006212; RTECS: DW1750000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C10H8N2; MW: 156.18
Melting Point: 70-73°; Boiling Point: 273°
- RTECS: DW1750000
- Crystalline
Formula:
C₁₀H₈N₂ Boiling Pt: 273 °C Melting Pt: 70…73 °C |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Assay | 98.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...