Lead(II) sulfide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Lead sulfide, Lead sulphide

127945-10G 127945-50G
BT127945-10GEA 68 USD
BT127945-10G BT127945-50G
Lead(II) sulfide ≥99.9% (trace metals basis)
Lead (II) sulphide

CAS: 1314-87-0; EC No: 215-246-6; MDL No: MFCD00016280; RTECS: OG4550000 UN No: UN3077; Haz Class: 9; Packing Group: III -200 Mesh Powder; Molecular Formula: PbS; MW: 239.25 Density (g/mL): 7.7


  • RTECS: OG4550000
  • -200 Mesh Powder

Formula: ̍PbS
MW: 239.27 g/mol
Boiling Pt: 1344
Melting Pt: 1114
Density: 7.7 g/mL
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9 III
Merck Index: 12,05445

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR