Quinidine
Supplier: BeanTown Chemical
Synonyms:
(+)-Quinidine
127895-1G
127895-25G
127895-5G
BT127895-1GEA
25
USD
BT127895-1G
BT127895-25G
BT127895-5G
Quinidine
Quinidine
CAS: 56-54-2; EC No: 200-279-0; MDL No: MFCD00135581; RTECS: VA4725000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C20H24N2O2; MW: 324.42
Melting Point: 168-172°
Light Sensitive
- RTECS: VA4725000
- Light Sensitive
- Crystalline
Formula:
C20H24N2O2 MW: 324.42 g/mol Melting Pt: 168…172 °C |
MDL Number:
MFCD00135581 CAS Number: 56-54-2 EINECS: 200-279-0 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...