N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99%
Supplier: BeanTown Chemical
CAS: 110-18-9; EC No: 203-744-6; MDL No: MFCD00008335; RTECS: KV7175000
UN No: UN2372; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2NCH2CH2N(CH3)2; Molecular Formula: C6H16N2; MW: 116.21
Melting Point: -55°; Boiling Point: 120-122°; Flash point: 18°C (62°F)
Density (g/mL): 0.775; Refractive Index: 1.418
Air Sensitive, Moisture Sensitive
- RTECS: KV7175000
- Refractive index: 1.418
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₆H₁₆N₂ Boiling Pt: 120…122 °C Melting Pt: –55 °C Density: 0.775 g/mL Flash Pt: 18 °C (62 °F) |
MDL Number:
MFCD00008335 CAS Number: 110-18-9 EINECS: 203-744-6 UN: 2372 ADR: 3,II |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...