Fluorescein disodium salt

Supplier: BeanTown Chemical

127830-100G 127830-2.5KG 127830-500G
BT127830-100GEA 30.48 USD
BT127830-100G BT127830-2.5KG BT127830-500G
Fluorescein disodium salt
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR