Fluorescein disodium salt
Supplier: BeanTown Chemical
127830-100G
127830-2.5KG
127830-500G
BT127830-100GEA
30.48
USD
BT127830-100G
BT127830-2.5KG
BT127830-500G
Fluorescein disodium salt
Fluorescein disodium salt
CAS: 518-47-8; EC No: 208-253-0; MDL No: MFCD00167039; RTECS: LM5425000
Powder; Molecular Formula: C20H10Na2O5; MW: 376.28
- RTECS: LM5425000
- Powder
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...