[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
[1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd(dppf)Cl2, 1,1’-Bis(diphenylphosphino)ferrocene palladium(II)dichloride
127795-1G
127795-5G
BT127795-1GEA
65.95
USD
BT127795-1G
BT127795-5G
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride ≥99%
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride
CAS: 72287-26-4; MDL No: MFCD00015757
Solid; Molecular Formula: C34H28FeP2·PdCl2; MW: 731.70
Melting Point: 266-283°
- Solid
Formula:
C₃₄H₂₈Cl₂P₂FePd Melting Pt: 266…283 °C |
MDL Number:
MFCD00015757 CAS Number: 72287-26-4 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...