2-Oxazolidinone ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Oxazolidone, Oxazolidin-2-one
127545-100G
127545-25G
127545-500G
BT127545-100GEA
119
USD
BT127545-100G
BT127545-25G
BT127545-500G
2-Oxazolidinone ≥99%
2-Oxazolidinone
CAS: 497-25-6; EC No: 207-840-9; MDL No: MFCD00005268; RTECS: RQ2450000
Crystalline; Molecular Formula: C3H5NO2; MW: 87.08
Melting Point: 83-87°; Boiling Point: 220°/48 mmHg
- RTECS: RQ2450000
- Crystalline
Formula:
C₃H₅NO₂ MW: 87.08 g/mol Boiling Pt: 220 °C/48 mmHg Melting Pt: 83…87 °C |
MDL Number:
MFCD00005268 CAS Number: 497-25-6 EINECS: 207-840-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...