2-Oxazolidinone ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Oxazolidone, Oxazolidin-2-one

127545-100G 127545-25G 127545-500G
BT127545-100GEA 119 USD
BT127545-100G BT127545-25G BT127545-500G
2-Oxazolidinone ≥99%
2-Oxazolidinone

CAS: 497-25-6; EC No: 207-840-9; MDL No: MFCD00005268; RTECS: RQ2450000 Crystalline; Molecular Formula: C3H5NO2; MW: 87.08 Melting Point: 83-87°; Boiling Point: 220°/48 mmHg


  • RTECS: RQ2450000
  • Crystalline

Formula: C₃H₅NO₂
MW: 87.08 g/mol
Boiling Pt: 220 °C/48 mmHg
Melting Pt: 83…87 °C
MDL Number: MFCD00005268
CAS Number: 497-25-6
EINECS: 207-840-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR