5-Amino-3-methylisoxazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

127525-1G 127525-25G 127525-5G
BT127525-1GEA 46 USD
BT127525-1G BT127525-25G BT127525-5G
5-Amino-3-methylisoxazole ≥98%
5-Amino-3-methylisoxazole

CAS: 14678-02-5; EC No: 238-719-9; MDL No: MFCD00003151 Solid; Molecular Formula: C4H6N2O; MW: 98.10 Melting Point: 85-87°


  • Solid

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 84
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR