Silicon carbide, fine powder 320 grit

Supplier: BeanTown Chemical

127430-10KG 127430-2.5KG 127430-500G
BT127430-10KGEA 319.9 USD
BT127430-10KG BT127430-2.5KG BT127430-500G
Silicon carbide, fine powder 320 grit
Silicon carbide

CAS: 409-21-2; EC No: 206-991-8; MDL No: MFCD00049531 Powder; Linear Formula: SiC; MW: 40.10 Melting Point: 2700° Density (g/mL): 3.22


  • Powder

Formula: SiC
Melting Pt: 2700 °C
Density: 3.22 g/mL
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR