Iron(III) oxide ≥98%

Supplier: BeanTown Chemical

Synonyms: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

127425-1KG 127425-5KG
BT127425-1KGEA 57.26 USD
BT127425-1KG BT127425-5KG
Iron(III) oxide ≥98%
Iron (III) oxide

CAS: 1309-37-1; EC No: 215-168-2; MDL No: MFCD00011008; RTECS: NO7400000 -325 Mesh Powder; Molecular Formula: Fe2O3 ; MW: 159.69


  • RTECS: NO7400000
  • -325 Mesh Powder

Formula: Fe₂O₃
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR