Sodium chlorite ≥80%, Technical Grade

Supplier: BeanTown Chemical

127350-10KG 127350-2KG 127350-500G
BT127350-10KGEA 257.87 USD
BT127350-10KG BT127350-2KG BT127350-500G
Sodium chlorite ≥80%, Technical Grade
Sodium chlorite

CAS: 7758-19-2; EC No: 231-836-6; MDL No: MFCD00003478; RTECS: VZ4800000 UN No: UN1496; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: NaClO2; MW: 90.44 Melting Point: 180-200° (decomposes) Hygroscopic


  • RTECS: VZ4800000
  • Hygroscopic
  • Powder

Formula: ClNaO₂
MW: 90.44 g/mol
Boiling Pt: 112
Melting Pt: 180…200 °C (dec.)
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1 II
Merck Index: 13,08672

Order Now

Specification Test Results

Assay 80%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR