Methyl cinnamate ≥99%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate

127305-100G 127305-2.5KG 127305-500G
BT127305-100GEA 26 USD
BT127305-100G BT127305-2.5KG BT127305-500G
Methyl cinnamate ≥99%
Methyl cinnamate

CAS: 103-26-4; EC No: 203-093-8; MDL No: MFCD00008458; RTECS: GE0190000 Crystalline; Linear Formula: C6H5CH=CHCOOCH3; Molecular Formula: C10H10O2; MW: 162.19 Melting Point: 33-38°; Boiling Point: 260-262°; Flash point: 110°C (230°F) Density (g/mL): 1.092


  • RTECS: GE0190000
  • Crystalline

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 260…262 °C
Melting Pt: 33…38 °C
Density: 1.092 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008458
CAS Number: 103-26-4
EINECS: 203-093-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR