Methyl cinnamate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate
127305-100G
127305-2.5KG
127305-500G
BT127305-100GEA
26
USD
BT127305-100G
BT127305-2.5KG
BT127305-500G
Methyl cinnamate ≥99%
Methyl cinnamate
CAS: 103-26-4; EC No: 203-093-8; MDL No: MFCD00008458; RTECS: GE0190000
Crystalline; Linear Formula: C6H5CH=CHCOOCH3; Molecular Formula: C10H10O2; MW: 162.19
Melting Point: 33-38°; Boiling Point: 260-262°; Flash point:
- RTECS: GE0190000
- Crystalline
Formula:
C₁₀H₁₀O₂ MW: 162.19 g/mol Boiling Pt: 260…262 °C Melting Pt: 33…38 °C Density: 1.092 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00008458 CAS Number: 103-26-4 EINECS: 203-093-8 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...