2,2'-Bipyridyl ≥99%
Supplier: BeanTown Chemical
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
127215-100G
127215-25G
127215-500G
BT127215-100GEA
173
USD
BT127215-100G
BT127215-25G
BT127215-500G
2,2'-Bipyridyl ≥99%
2,2'-Bipyridyl
CAS: 366-18-7; EC No: 206-674-4; MDL No: MFCD00006212; RTECS: DW1750000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C10H8N2; MW: 156.18
Melting Point: 70-73°; Boiling Point: 273°
- RTECS: DW1750000
- Solid
Formula:
C₁₀H₈N₂ Boiling Pt: 273 °C Melting Pt: 70…73 °C |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...