Silver conductive adhesive paste

Supplier: BeanTown Chemical

127185-15G
BT127185-15GEA 255.62 USD
BT127185-15G
Silver conductive adhesive paste
Silver conductive adhesive paste

CAS: 7440-22-4; EC No: 232-033-3; MDL No: MFCD01868500 Paste; Molecular Formula: Ag; MW: 107.87 Boiling Point: 80°; Flash point: 110°C (230°F)


  • Paste

Boiling Pt: 80 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD01868500
EINECS: 232-033-3

Order Now

Specification Test Results

Sheet resistance <0.025ohm/square @ 0.001in thick

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR