4-Nitro-2,3-lutidine-N-oxide ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,3-Dimethyl-4-nitropyridine 1-oxide, 2,3-Dimethyl-4-nitropyridine N-oxide
127180-1G
127180-25G
127180-5G
BT127180-1GEA
29
USD
BT127180-1G
BT127180-25G
BT127180-5G
4-Nitro-2,3-lutidine-N-oxide ≥97%
4-Nitro-2,3-lutidine N-oxide
CAS: 37699-43-7; MDL No: MFCD00065172; RTECS: UT2807000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H8N2O3; MW: 168.15
Melting Point: 94-98°
- RTECS: UT2807000
- Solid
Formula:
C₇H₈N₂O₂ Melting Pt: 94…98 °C |
MDL Number:
MFCD00065172 CAS Number: 37699-43-7 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...