Cesium hydroxide 50% (w/w) in water , 99% (trace metals basis)

Supplier: BeanTown Chemical

127130-C100G 127130-C500G
BT127130-C100GEA 116.09 USD
BT127130-C100G BT127130-C500G
Cesium hydroxide 50% (w/w) in water , 99% (trace metals basis)
Cesium hydroxide

CAS: 21351-79-1; EC No: 244-344-1; MDL No: MFCD00010964 UN No: UN2681; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CsOH; MW: 149.91 Density (g/mL): 1.72 Air Sensitive


  • Air Sensitive
  • Liquid

Formula: CsOH
Density: 1.72 g/mL
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR