Aluminium ≥99.999% (trace metals basis), slug, Length 6.35 mm, Diameter 6.35 mm

Supplier: BeanTown Chemical

Synonyms: Aluminum

127105-10G 127105-250G 127105-50G
BT127105-10GEA 27 USD
BT127105-10G BT127105-250G BT127105-50G
Aluminium ≥99.999% (trace metals basis), slug, Length 6.35 mm, Diameter 6.35 mm
Aluminium

CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330002 Solid; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7


  • RTECS: BD0330002
  • Solid

Formula: Al
Boiling Pt: 2460 °C
Melting Pt: 660.4 °C
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR