Neodymium ≥99.5% (trace metals basis excluding Ta), rods Ø 6.35 mm
Supplier: BeanTown Chemical
127035-100MM
127035-10MM
127035-50MM
BT127035-100MMEA
507
USD
BT127035-100MM
BT127035-10MM
BT127035-50MM
Neodymium ≥99.5% (trace metals basis excluding Ta), rods Ø 6.35 mm
Neodymium
CAS: 7440-00-8; EC No: 231-109-3; MDL No: MFCD00011130; RTECS: QO8575000
Solid, cast rod, diameter is approximate; Molecular Formula: Nd; MW: 144.24
Melting Point: 1021°; Boiling Point: 3074°
Density (g/mL): 7.003
- RTECS: QO8575000
- Solid, cast rod, diameter is approximate
Formula:
Nd Boiling Pt: 3074 °C Melting Pt: 1021 °C Density: 7.003 g/mL |
MDL Number:
MFCD00011130 CAS Number: 7440-00-8 EINECS: 231-109-3 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...