Barium carbonate ≥99.995%

Supplier: BeanTown Chemical

126925-100G 126925-25G 126925-500G
BT126925-100GEA 332 USD
BT126925-100G BT126925-25G BT126925-500G
Barium carbonate ≥99.995%
Barium carbonate

CAS: 513-77-9; EC No: 208-167-3; MDL No: MFCD00003448; RTECS: CQ8600000 -22 Mesh Powder; Linear Formula: BaCO3; MW: 197.35


  • RTECS: CQ8600000
  • -22 Mesh Powder

Formula: BaCO₃
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR