Methyl arachidate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyl eicosanoate, Methyl Arachidate
126870-1G
126870-5G
BT126870-1GEA
29.94
USD
BT126870-1G
BT126870-5G
Methyl arachidate ≥98%
Methyl arachidate
CAS: 1120-28-1; EC No: 214-304-8; MDL No: MFCD00009014
Solid; Linear Formula: CH3(CH2)18COOCH3; Molecular Formula: C21H42O2; MW: 326.56
Melting Point: 45-48°; Boiling Point: 215°/10 mmHg; Flash point:
- Solid
Formula:
C₂₁H₄₂O₂ Boiling Pt: 215 °C/10 mmHg Melting Pt: 45…48 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00009014 CAS Number: 1120-28-1 EINECS: 214-304-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...