Silver ≥99.9% (trace metals basis), nanopowder
Supplier: BeanTown Chemical
CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000
UN No: UN3077; Haz Class: 9; Packing Group: III
20-40nm APS Powder; Molecular Formula: Ag; MW: 107.87
Melting Point: 960°; Boiling Point: 2212°; Flash point: 111°C (232°F)
Density (g/mL): 10.49
Air Sensitive
- RTECS: VW3500000
- Air Sensitive
- 20-40nm APS Powder
Formula:
Ag Boiling Pt: 2212 °C Melting Pt: 960 °C Density: 10.49 g/mL Flash Pt: 111 °C (232 °F) |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 ADR: 9,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...