Isobutyl chloroformate ≥98%

Supplier: BeanTown Chemical

126750-100G 126750-25G 126750-500G
BT126750-100GEA 37.89 USD
BT126750-100G BT126750-25G BT126750-500G
Isobutyl chloroformate ≥98%
Isobutyl chloroformate

CAS: 543-27-1; EC No: 208-840-1; MDL No: MFCD00000642 UN No: UN2742; Haz Class: 6.1 (8,3); Packing Group: II Liquid; Molecular Formula: C5H9ClO2; MW: 136.58 Boiling Point: 128-129°; Flash point: 30°C (86°F) Density (g/mL): 1.053; Refractive Index: 1.406 Moisture Sensitive


  • Refractive index: 1.406
  • Moisture Sensitive
  • Liquid

Formula: C₅H₉ClO₂
Boiling Pt: 128…129 °C
Density: 1.053 g/mL
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD00000642
CAS Number: 543-27-1
EINECS: 208-840-1
UN: 2742
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR