Yttrium aluminum oxide ≥99.9%

Supplier: BeanTown Chemical

126710-100G 126710-25G
BT126710-100GEA 709 USD
BT126710-100G BT126710-25G
Yttrium aluminum oxide ≥99.9%
Yttrium aluminum oxide

CAS: 12005-21-9; EC No: 234-465-8; MDL No: MFCD00213958 Powder; Linear Formula: Y3Al5O12; MW: 593.62 Melting Point: 1760° Density (g/mL): 4.56; Refractive Index: 1.833


  • Refractive index: 1.833
  • Powder

Formula: Y₃Al₅O₁₂
MW: 593.62 g/mol
Melting Pt: 1760 °C
Density: 4.56 g/mL
MDL Number: MFCD00213958
CAS Number: 12005-21-9
EINECS: 234-465-8

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR