Glycine ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

126695-1KG 126695-250G
BT126695-1KGEA 113.97 USD
BT126695-1KG BT126695-250G
Glycine ≥99.5%
Glycine

CAS: 56-40-6; EC No: 200-272-2; MDL No: MFCD00008131; RTECS: MB7600000 Powder; Linear Formula: NH2CH2CO2H; MW: 75.07 Melting Point: 240° (decomposes)


  • RTECS: MB7600000
  • Powder

Formula: NH₂CH₂COOH
Melting Pt: 240 °C (dec.)
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR