5-Acetylvaleric acid ≥96%

Supplier: BeanTown Chemical

Synonyms: 6-Oxoheptanoic acid

126640-10G 126640-50G
BT126640-10GEA 112 USD
BT126640-10G BT126640-50G
5-Acetylvaleric acid ≥96%
5-Acetylvaleric acid

CAS: 3128-07-2; EC No: 221-512-2; MDL No: MFCD00051479 UN No: UN3261; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C7H12O3; MW: 144.17 Melting Point: 35-37°; Boiling Point: 158-162°/9 mmHg Density (g/mL): 1.059


  • Solid

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 158…162 °C/9 mmHg
Melting Pt: 35…37 °C
Density: 1.059 g/mL
MDL Number: MFCD00051479
CAS Number: 3128-07-2
EINECS: 221-512-2
ADR: 8 III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR