Dimethyl acetylenedicarboxylate ≥95%
Supplier: BeanTown Chemical
126555-100G
126555-25G
126555-500G
BT126555-100GEA
181
USD
BT126555-100G
BT126555-25G
BT126555-500G
Dimethyl acetylenedicarboxylate ≥95%
Dimethyl acetylenedicarboxylate
CAS: 762-42-5; EC No: 212-098-4; MDL No: MFCD00008456; RTECS: ES0175000
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: CH3O2CC≡CCO2CH3; Molecular Formula: C6H6O4; MW: 142.11
Boiling Point: 95-98°/19 mmHg; Flash point: 86°C (187°F)
Density (g/mL): 1.156; Refractive Index: 1.447
Light Sensitive
- RTECS: ES0175000
- Refractive index: 1.447
- Light Sensitive
- Liquid
Formula:
C₆H₆O₄ Boiling Pt: 95…98 °C/19 mmHg Density: 1.156 g/mL Flash Pt: 86 °C (187 °F) |
MDL Number:
MFCD00008456 CAS Number: 762-42-5 EINECS: 212-098-4 ADR: 8,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...