1,3-Dibromoadamantane ≥99%
Supplier: BeanTown Chemical
126535-1G
BT126535-1GEA
80
USD
BT126535-1G
1,3-Dibromoadamantane ≥99%
1,3-Dibromoadamantane
CAS: 876-53-9; MDL No: MFCD00077218
Solid; Molecular Formula: C10H14Br2; MW: 294.03
Melting Point: 108-110°
- Solid
Formula:
C₁₀H₁₄Br₂ Melting Pt: 108…110 °C |
MDL Number:
MFCD00077218 CAS Number: 876-53-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...