Trypan blue
Supplier: BeanTown Chemical
126515-100G
126515-25G
126515-500G
BT126515-100GEA
121.48
USD
BT126515-100G
BT126515-25G
BT126515-500G
Trypan blue
Trypan Blue
CAS: 72-57-1; EC No: 200-786-7; MDL No: MFCD00003969; RTECS: QJ6475000
Solid; Molecular Formula: C34H24N6O14S4Na4; MW: 960.82
Melting Point:
- RTECS: QJ6475000
- Solid
Formula:
C₃₄H₂₄N₆Na₄O₁₄S₄ Melting Pt: >300 °C |
MDL Number:
MFCD00003969 CAS Number: 72-57-1 EINECS: 200-786-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...