n-Pentacosane ≥99%

Supplier: BeanTown Chemical

126375-1G 126375-25G 126375-5G
BT126375-1GEA 58 USD
BT126375-1G BT126375-25G BT126375-5G
n-Pentacosane ≥99%
n-Pentacosane

CAS: 629-99-2; EC No: 211-123-6; MDL No: MFCD00009353 Solid; Linear Formula: CH3(CH2)23CH3; Molecular Formula: C25H52; MW: 352.67 Melting Point: 53-56°; Boiling Point: 169-170°/0.05 mmHg; Flash point: 120°C (248°F) Density (g/mL): 0.779; Refractive Index: 1.438


  • Refractive index: 1.438
  • Solid

Formula: H₃C(CH₂)₂₃CH₃
Boiling Pt: 169…170 °C/0.05 mmHg
Melting Pt: 53…56 °C
Density: 0.779 g/mL
Flash Pt: 120 °C (248 °F)
MDL Number: MFCD00009353
CAS Number: 629-99-2
EINECS: 211-123-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR