2-Bromopyridine ≥99%
Supplier: BeanTown Chemical
126350-100G
126350-250G
126350-25G
BT126350-100GEA
86
USD
BT126350-100G
BT126350-250G
BT126350-25G
2-Bromopyridine ≥99%
2-Bromopyridine
CAS: 109-04-6; EC No: 203-641-6; MDL No: MFCD00006219; RTECS: US3850000
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C5H4BrN; MW: 158.00
Boiling Point: 192-194°
Density (g/mL): 1.657; Refractive Index: 1.572
- RTECS: US3850000
- Refractive index: 1.572
- Liquid
Formula:
C₅H₄BrN Boiling Pt: 192…194 °C Density: 1.657 g/mL |
MDL Number:
MFCD00006219 CAS Number: 109-04-6 EINECS: 203-641-6 UN: 2810 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...