Aluminium oxide aerosol refractory paint

Supplier: BeanTown Chemical

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

126120-12OZ 126120-5X12OZ
BT126120-12OZEA 144 USD
BT126120-12OZ BT126120-5X12
Aluminium oxide aerosol refractory paint
Aluminium oxide

CAS: 1344-28-1; MDL No: MFCD00003424 UN No: UN1950; Haz Class: 2.1 Aerosol Spray Can; Molecular Formula: Al2O3; MW: 101.96


  • Aerosol Spray Can

Formula: Al₂O₃
MDL Number: MFCD00003424
CAS Number: 1344-28-1
ADR: 2.1,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR