Gallium ≥99.99999% (trace metals basis), pellets, Diameter 6 mm

Supplier: BeanTown Chemical

126015-10G 126015-1G 126015-50G
BT126015-10GEA 100 USD
BT126015-10G BT126015-1G BT126015-50G
Gallium ≥99.99999% (trace metals basis), pellets, Diameter 6 mm
Gallium
Formula: Ga
Boiling Pt: 2403 °C
Melting Pt: 29.8 °C
Density: 5.904 g/mL
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III

Order Now

Specification Test Results

Assay 99.99999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR