Tetraethylene glycol monomethyl ether ≥98%

Supplier: BeanTown Chemical

Synonyms: mPEG4-OH, 2,5,8,11-Tetraoxatridecan-13-ol

126010-25G 126010-5G
BT126010-25GEA 282 USD
BT126010-25G BT126010-5G
Tetraethylene glycol monomethyl ether ≥98%
Tetraethylene glycol monomethyl ether

CAS: 23783-42-8; EC No: 245-883-5; MDL No: MFCD00041756 Liquid; Linear Formula: CH3(OCH2CH2)4OH; Molecular Formula: C9H20O5; MW: 208.25 Melting Point: -39°; Boiling Point: 163-166°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.045; Refractive Index: 1.444


  • Refractive index: 1.444
  • Liquid

Formula: C₉H₂₀O₅
MW: 208.25 g/mol
Boiling Pt: 163…166 °C/11 mmHg
Melting Pt: –39 °C
Density: 1.045 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00041756
CAS Number: 23783-42-8
EINECS: 245-883-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR