Tributyltin fluoride

Supplier: BeanTown Chemical

Synonyms: Tri-n-butyltin fluoride, Fluorotributylstannane

125955-25G 125955-5G
BT125955-25GEA 161 USD
BT125955-25G BT125955-5G
Tributyltin fluoride
Tributyltin fluoride

CAS: 1983-10-4; EC No: 217-847-9; MDL No: MFCD00013547 UN No: UN3146; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: [CH3(CH2)3]3SnF; Molecular Formula: C12H27FSn; MW: 309.04 Melting Point: 248-252° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₁₂H₂₇FSn
MW: 309.05 g/mol
Melting Pt: 248…252 °C
MDL Number: MFCD00013547
CAS Number: 1983-10-4
EINECS: 217-847-9
UN: 3146
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR