2-Picoline-N-oxide ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Methylpyridine-N-oxide, 2-Picoline N-oxide, 2-Methylpyridine-1-oxide, 2-Methylpyridine 1-oxide

125880-100G 125880-25G 125880-500G
BT125880-100GEA 71 USD
BT125880-100G BT125880-25G BT125880-500G
2-Picoline-N-oxide ≥98%
2-Picoline N-oxide

CAS: 931-19-1; EC No: 213-230-3; MDL No: MFCD00006199 Crystalline; Molecular Formula: C6H7NO; MW: 109.13 Melting Point: 41-45°; Flash point: 143°C (289°F) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 259
Melting Pt: 0
Flash Pt: 143 °C (289 °F)
MDL Number: MFCD00006199
CAS Number: 931-19-1
EINECS: 213-230-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR