Disodium-2,6-naphthalenedisulfonate ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,6-Naphthalenedisulfonic acid disodium salt, Sodium naphthalene-2,6-disulfonate, Sodium naphthalene-2,6-disulphonate, Ebert-Merz beta-acid disodium salt, 2,6-Naphthalenedisulphonic acid disodium salt
125795-100G
BT125795-100GEA
100
USD
BT125795-100G
Disodium-2,6-naphthalenedisulfonate ≥97%
Disodium-2,6-naphthalenedisulfonate
CAS: 1655-45-4; EC No: 216-735-7; MDL No: MFCD00004092
Solid; Linear Formula: C10H6(SO3Na)2; MW: 332.26
Melting Point:
- Solid
Formula:
C₁₀H₈O₆S₂ Melting Pt: >300 °C |
MDL Number:
MFCD00004092 CAS Number: 1655-45-4 EINECS: 216-735-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...