4-Amino-1-(2-pyridyl)piperidine hydrochloride ≥97%

Supplier: BeanTown Chemical

125760-1G 125760-250MG
BT125760-1GEA 304 USD
BT125760-1G BT125760-250MG
4-Amino-1-(2-pyridyl)piperidine hydrochloride ≥97%
4-Amino-1-(2-pyridyl)piperidine hydrochloride

CAS: 77145-39-2; MDL No: MFCD09607833 Solid; Molecular Formula: C10H15N3·HCl ; MW: 213.71


  • Solid

Formula: C₁₀H₁₆ClN₃
MDL Number: MFCD09607833
CAS Number: 77145-39-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR