Cobalt(II,III) oxide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

125740-1KG 125740-250G 125740-50G
BT125740-1KGEA 750 USD
BT125740-1KG BT125740-250G BT125740-50G
Cobalt(II,III) oxide ≥99.5% (trace metals basis)
Cobalt (II,III) oxide

CAS: 1308-06-1; EC No: 215-157-2; MDL No: MFCD00010939; RTECS: GG2500000 UN No: UN3077; Haz Class: 9; Packing Group: III 2-6 Micron Powder; Molecular Formula: Co3O4; MW: 240.80 Melting Point: 895° (decomposes) Density (g/mL): 6.11


  • RTECS: GG2500000
  • 2-6 Micron Powder

Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 895 (dec.)
Density: 6.11 g/mL
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
ADR: 9 III
Merck Index: 13,02454

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR