4-Amino-5-bromo-2-chloropyridine ≥95%
Supplier: BeanTown Chemical
Synonyms:
5-bromo-2-chloropyridin-4-amine, 5-Bromo-2-chloro-4-pyridylamine, 5-Bromo-2-chloro-4-pyridinamine
125710-1G
125710-250MG
BT125710-1GEA
427
USD
BT125710-1G
BT125710-250MG
4-Amino-5-bromo-2-chloropyridine ≥95%
4-Amino-5-bromo-2-chloropyridine
CAS: 857730-21-3; MDL No: MFCD09037459
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C5H4BrClN2; MW: 207.46
Melting Point: 127-128°
- Solid
Formula:
C₅H₄BrClN₂ Melting Pt: 127…128 °C |
MDL Number:
MFCD09037459 CAS Number: 857730-21-3 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...