Formamide ≥99.0% for spectrophotometry

Supplier: BeanTown Chemical

Synonyms: Carbamaldehyde

125695-100ML 125695-4X500ML 125695-500ML
BT125695-100MLEA 38 USD
BT125695-100ML BT125695-4X500 BT125695-500ML
Formamide ≥99.0% for spectrophotometry
Formamide

CAS: 75-12-7; EC No: 200-842-0; MDL No: MFCD00007941; RTECS: LQ0525000 Liquid ; Linear Formula: HCONH2; MW: 45.04 Melting Point: 2-3°; Boiling Point: 210°; Flash point: 175°C (347°F) Density (g/mL): 1.134; Refractive Index: 1.447 Hygroscopic


  • RTECS: LQ0525000
  • Refractive index: 1.447
  • Hygroscopic
  • Liquid

Formula: HCONH₂
Boiling Pt: 210 °C
Melting Pt: 2…3 °C
Density: 1.134 g/mL
Flash Pt: 175 °C (347 °F)
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR