Formamide ≥99.0% for spectrophotometry
Supplier: BeanTown Chemical
Synonyms:
Carbamaldehyde
125695-100ML
125695-4X500ML
125695-500ML
BT125695-100MLEA
38
USD
BT125695-100ML
BT125695-4X500
BT125695-500ML
Formamide ≥99.0% for spectrophotometry
Formamide
CAS: 75-12-7; EC No: 200-842-0; MDL No: MFCD00007941; RTECS: LQ0525000
Liquid ; Linear Formula: HCONH2; MW: 45.04
Melting Point: 2-3°; Boiling Point: 210°; Flash point: 175°C (347°F)
Density (g/mL): 1.134; Refractive Index: 1.447
Hygroscopic
- RTECS: LQ0525000
- Refractive index: 1.447
- Hygroscopic
- Liquid
Formula:
HCONH₂ Boiling Pt: 210 °C Melting Pt: 2…3 °C Density: 1.134 g/mL Flash Pt: 175 °C (347 °F) |
MDL Number:
MFCD00007941 CAS Number: 75-12-7 EINECS: 200-842-0 |
|
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...