Potassium sulfate ≥99.995%
Supplier: BeanTown Chemical
125630-25G
125630-5G
BT125630-25GEA
176
USD
BT125630-25G
BT125630-5G
Potassium sulfate ≥99.995%
Potassium sulphate
CAS: 7778-80-5; EC No: 231-915-5; MDL No: MFCD00011388; RTECS: TT5900000
Powder; Linear Formula: K2SO4; MW: 174.26
Melting Point: 1069°; Boiling Point: 1689°
Density (g/mL): 2.662
- RTECS: TT5900000
- Powder
Formula:
K₂SO₄ Boiling Pt: 1689 °C Melting Pt: 1069 °C Density: 2.662 g/mL |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...