Yttrium(III) acetylacetonate hydrate ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Yttrium (III) tris(2,4-pentanedionato-O,O') hydrate, Yttrium (III) 2,4-pentanedionate hydrate, Tris(2,4-pentanedionato-O,O')yttrium (III) hydrate
125595-100G
125595-25G
BT125595-100GEA
413
USD
BT125595-100G
BT125595-25G
Yttrium(III) acetylacetonate hydrate ≥99.9% (trace rare earth metals basis)
Yttrium (III) acetylacetonate hydrate
CAS: 207801-29-4; EC No: 239-607-2; MDL No: MFCD00013509
Powder; Linear Formula: Y(C5H7O2)3·xH2O; Molecular Formula: C15H21O6Y·xH2O; MW: 386.24 (anhydrous)
- Powder
Formula:
C₁₅H₂₁O₆Y·H₂O MW: 404.25 |
MDL Number:
MFCD00013509 CAS Number: 207801-29-4 EINECS: 239-607-2 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...