Lithium carbonate ≥99%
Supplier: BeanTown Chemical
125545-2KG
125545-500G
BT125545-2KGEA
863.76
USD
BT125545-2KG
BT125545-500G
Lithium carbonate ≥99%
Lithium carbonate
CAS: 554-13-2; EC No: 209-062-5; MDL No: MFCD00011084
Powder; Linear Formula: Li2CO3; MW: 73.90
Melting Point: 618°
- Powder
Formula:
Li₂CO₃ MW: 73.89 g/mol Melting Pt: 720 |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 13,05549 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...